Learn More
KCNV2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21756-1-AP-20UL
This item is not returnable.
View return policy
Description
The KCNV2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KCNV2. This antibody recognizes human, mouse antigen. The KCNV2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| KCNV2 | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| KCNV2, KV11.1, Kv8.2, RCD3B | |
| 20 μL | |
| 169522 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |