missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNV2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101770
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KCNV2.
KCNV2 is a potassium channel subunit. Modulates channel activity by shifting the threshold and the half-maximal activation to more negative values.
Specifications
| KCNV2 | |
| Polyclonal | |
| Unconjugated | |
| KCNV2 | |
| Kcnv2; KV11.1; Kv8.2; potassium channel, subfamily V, member 2; potassium channel, voltage gated modifier subfamily V, member 2; potassium channel, voltage-gated modifier subfamily V, member 2; potassium voltage-gated channel modifier subfamily V member 2; potassium voltage-gated channel subfamily V member 2; RCD3B; voltage-gated potassium channel subunit Kv8.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 169522, 240595 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8CFS6, Q8TDN2 | |
| KCNV2 | |
| A synthesized peptide derived from human KCNV2(Accession Q8TDN2), corresponding to amino acid residues M1-Q40. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction