missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: GIYRTESQKL TTSESQISIS VEEWEDTKDS KEQGHHRSNH RNSTSSDQSD HPLLRR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Kcnt2 calcium (Ca2+) -activated potassium channels (KCa) are a group of 6/7-TM ion channels that selectively transport K+ ions across biological membranes. KCa channels are broadly classified into three subtypes: SK, IK and BK channels (small, intermediate and big conductance). Kcnt2 is an outward rectifying potassium channel, and produces rapidly activating outward rectifier K(+) currents. Kcnt2 is activated by high intracellular sodium and chloride levels. Further, Kcnt2 channel activity is inhibited by ATP and by inhalation anesthetics, such as isoflurane. Functionally, Kcnt2 is inhibited upon stimulation of G-protein coupled receptors, such as CHRM1 and GRIA1.
Specifications
Specifications
| Antigen | KCNT2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Kcnt2 |
| Gene Accession No. | Q6UVM3 |
| Gene Alias | E330038N15Rik; KCa4.2; Kcnt2; Potassium channel subfamily T member 2; potassium channel, sodium activated subfamily T, member 2; potassium channel, sodium-activated subfamily T, member 2; potassium channel, subfamily T, member 2; potassium sodium-activated channel subfamily T member 2; sequence like an intermediate conductance potassium channel subunit; SLICK; SLO2.1; sodium- and chloride-activated ATP-sensitive potassium channel; sodium and chloride-activated ATP-sensitive potassium channel Slo2.1; sodium- and chloride-activated ATP-sensitive potassium channel subunit; sodium-and chloride-activated ATP-sensitive potassium channel (SLICK) |
| Gene Symbols | Kcnt2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?