missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577638
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a sodium-activated potassium channel subunit which is thought to function in ion conductance and developmental signaling pathways. Mutations in this gene cause the early-onset epileptic disorders, maligt migrating partial seizures of infancy and autosomal domit nocturnal frontal lobe epilepsy. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Dec 2012].
Specifications
| KCNT1 | |
| Polyclonal | |
| Unconjugated | |
| Kcnt1 | |
| Slo2.2, KCNT1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 227632, 60444 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q6ZPR4, Q9Z258 | |
| Kcnt1 | |
| Peptide (C)KQEEKQNRRGLAG, corresponding to amino acid residues 619-631 of rat Slack | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction