missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNQ5 Polyclonal Antibody

Description
Immunogen sequence: LALASFQIPP FECEQTSDYQ SPVDSKDLSG SAQNSGCLSR STSANISRGL QFILTPNEFS AQTFYALSPT MHSQATQVPI SQSDGSAVAA TNTIANQINT APKPAAPTTL QIPPPL Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 91%.
KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KNCQs (KCNQ1-5) found in the central nervous system. Studies have shown that KCNQ3 and KCNQ5 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy. Voltage-dependent potassium channels are key regulators of the resting membrane potential and modulate the excitability of electrically active cells. The channels are usually tetrameric and can interact with auxiliary subunits that enhance or modify currents mediated by the pore-forming subunits.
Specifications
Specifications
| Antigen | KCNQ5 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KCNQ5 |
| Gene Accession No. | Q9NR82 |
| Gene Alias | 7730402H11; 9230107O05Rik; AA589396; D1Mgi1; KCNQ5; Kcnq5l; KQT-like 5; Kv7.5; KvLQT5; potassium channel protein; potassium channel subunit alpha KvLQT5; potassium channel, voltage gated KQT-like subfamily Q, member 5; potassium channel, voltage-gated KQT-like subfamily Q, member 5; potassium voltage-gated channel subfamily KQT member 5; potassium voltage-gated channel subfamily Q member 5; potassium voltage-gated channel, KQT-like subfamily, member 5; potassium voltage-gated channel, subfamily Q, member 5; rCG_43583; voltage-gated potassium channel subunit Kv7.5; voltage-gated potassium channel type Kv7.5 |
| Gene Symbols | KCNQ5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?