missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNQ5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 30019532
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019532 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30019532 Supplier Invitrogen™ Supplier No. PA5144594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KCNQ5 Polyclonal Antibody for Western Blot

KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KNCQs (KCNQ1-5) found in the central nervous system. Studies have shown that KCNQ3 and KCNQ5 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy. Voltage-dependent potassium channels are key regulators of the resting membrane potential and modulate the excitability of electrically active cells. The channels are usually tetrameric and can interact with auxiliary subunits that enhance or modify currents mediated by the pore-forming subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNQ5
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal
Gene KCNQ5
Gene Accession No. Q9NR82
Gene Alias 7730402H11; 9230107O05Rik; AA589396; D1Mgi1; KCNQ5; Kcnq5l; KQT-like 5; Kv7.5; KvLQT5; potassium channel protein; potassium channel subunit alpha KvLQT5; potassium channel, voltage gated KQT-like subfamily Q, member 5; potassium channel, voltage-gated KQT-like subfamily Q, member 5; potassium voltage-gated channel subfamily KQT member 5; potassium voltage-gated channel subfamily Q member 5; potassium voltage-gated channel, KQT-like subfamily, member 5; potassium voltage-gated channel, subfamily Q, member 5; rCG_43583; voltage-gated potassium channel subunit Kv7.5; voltage-gated potassium channel type Kv7.5
Gene Symbols KCNQ5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human KCNQ5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56479
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.