missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNQ4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577657
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
The protein encoded by this gene forms a potassium channel that is thought to play a critical role in the regulation of neuronal excitability, particularly in sensory cells of the cochlea. The current generated by this channel is inhibited by M1 muscarinic acetylcholine receptors and activated by retigabine, a novel anti-convulsant drug. The encoded protein can form a homomultimeric potassium channel or possibly a heteromultimeric channel in association with the protein encoded by the KCNQ3 gene. Defects in this gene are a cause of nonsyndromic sensorineural deafness type 2, an autosomal domit form of progressive hearing loss. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| KCNQ4 | |
| Polyclonal | |
| Unconjugated | |
| Kcnq4 | |
| KV7.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 298496, 60613 | |
| -20°C | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q9JK96, Q9JK97 | |
| Kcnq4 | |
| Peptide (C)DRKTREKGDKGPSD, corresponding to amino acid residues 594-607 of mouse KV 7.4 | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction