missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNN4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23271-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is part of a potentially heterotetrameric voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization, which promotes calcium influx. The encoded protein may be part of the predomit calcium-activated potassium channel in T-lymphocytes. This gene is similar to other KCNN family potassium channel genes, but it differs enough to possibly be considered as part of a new subfamily.Specifications
| KCNN4 | |
| Polyclonal | |
| Unconjugated | |
| KCNN4 | |
| KCa3.1, KCNN4, SK4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3783 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15554 | |
| KCNN4 | |
| KCNN4 Fusion Protein Ag19774 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction