missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNN4 Mouse anti-Human, Mouse, Rat, Clone: 2A6G5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | KCNN4 |
|---|---|
| Clone | 2A6G5 |
| Concentration | 1.05 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16887335
|
Proteintech
60276-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16877335
|
Proteintech
60276-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is part of a potentially heterotetrameric voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization, which promotes calcium influx. The encoded protein may be part of the predomit calcium-activated potassium channel in T-lymphocytes. This gene is similar to other KCNN family potassium channel genes, but it differs enough to possibly be considered as part of a new subfamily.Specifications
| KCNN4 | |
| 1.05 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hIKCa1, hKCa4, hSK4, IK1, IKCA1, KCa3.1, KCA4, KCNN4, Putative Gardos channel, SK4, SKCa 4, SKCa4 | |
| KCNN4 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 2A6G5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat, Human | |
| O15554, O89109, Q9QYW1 | |
| 16534, 3783, 65206 | |
| KCNN4 Fusion Protein Ag19774 | |
| Primary | |
| -20°C | |
| KCNN4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title