missing translation for 'onlineSavingsMsg'
Learn More

KCNN4 Antibody, Novus Biologicals™

Product Code. 18367088 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367088 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367088 Supplier Novus Biologicals Supplier No. H00003783B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

KCNN4 Polyclonal antibody specifically detects KCNN4 in Human, Porcine samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNN4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_002241.1
Gene Alias hIKCa1, hKCa4, hSK4, IK1, IKCa1, intermediate conductance calcium-activated potassium channel protein 4, KCa3.1IKCA1, KCa4, KCA4SKCa4, potassium intermediate/small conductance calcium-activated channel, subfamilyN, member 4, putative erythrocyte intermediate conductance calcium-activated potassiumGardos channel, Putative Gardos channel, SK4SKCa 4
Host Species Mouse
Immunogen KCNN4 (NP_002241.1, 1 a.a. - 427 a.a.) full-length human protein. MGGDLVLGLGALRRRKRLLEQEKSLAGWALVLAGTGIGLMVLHAEMLWFGGCSWALYLFLVKCTISISTFLLLCLIVAFHAKEVQLFMTDNGLRDWRVALTGRQAAQIVLELVVCGLHPAPVRGPPCVQDLGAPLTSPQPWPGFLGQGEALLSLAMLLRLYLVPRAVLLRSGVLLNASYRSIGALNQVRFRHWFVAKLYMNTHPGRLLLGLTLGLWLTTAWVLSVAERQAVNATGHLSDTLWLIPITFLTIGYGDVVPGTMWGKIVCLCTGVMGVCCTALLVAVVARKLEFNKAEKHVHNFMMDIQYTKEMKESAARVLQEAWMFYKHTRRKESHAARRHQRKLLAAINAFRQVRLKHRKLREQVNSMVDISKMHMILYDLQQNLSSSHRALEKQIDTLAGKLDALTELLSTALGPRQLPEPSQQSK
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3783
Target Species Human, Pig
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.