missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNN4 Antibody (8A11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42987
This item is not returnable.
View return policy
Description
KCNN4 Monoclonal antibody specifically detects KCNN4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| KCNN4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| 8A11 | |
| Western Blot 1:500 - 1:2000, ELISA | |
| hIKCa1, hKCa4, hSK4, IK1, IKCa1, intermediate conductance calcium-activated potassium channel protein 4, KCa3.1IKCA1, KCa4, KCA4SKCa4, potassium intermediate/small conductance calcium-activated channel, subfamilyN, member 4, putative erythrocyte intermediate conductance calcium-activated potassiumGardos channel, Putative Gardos channel, SK4SKCa 4 | |
| potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4 - (Uniprot# O15554) | |
| 100 μg | |
| Cell Biology, Neuroscience | |
| 3783 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction