missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566058
This item is not returnable.
View return policy
Description
Immunogen sequence: NLIEAETEGQP LQLFSPSNPP EIVISSREDN HAHQTLLHHP NATHNHQHAG TTASSTTFPK ANKRK Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.
Action potentials in vertebrate neurons are followed by an afterhyperpolarization (AHP) that may persist for several seconds and may have profound consequences for the firing pattern of the neuron. Each component of the AHP is kinetically distinct and is mediated by different calcium-activated potassium channels. KCNN3, a member of the KCNN family of potassium channel genes, encodes a protein that is activated before membrane hyperpolarization and is thought to regulate neuronal excitability by contributing to the slow component of synaptic AHP. The encoded protein is an integral membrane protein that forms a voltage-independent calcium-activated channel with three other calmodulin-binding subunits.
Specifications
| KCNN3 | |
| Polyclonal | |
| Unconjugated | |
| KCNN3 | |
| hSK3; K3; KCa2.3; Kcnn3; potassium calcium-activated channel subfamily N member 3; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 3; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 3; SK3; SKCa 3; SKCa3; small conductance calcium-activated potassium channel 3; small conductance calcium-activated potassium channel protein 3; small-conductance Ca2+-activated K+ channel 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3782 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UGI6 | |
| KCNN3 | |
| Recombinant Human KCNN3. Recombinant protein control fragment (Product #RP-106751). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction