missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNN3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33423-20ul
This item is not returnable.
View return policy
Description
KCNN3 Monoclonal antibody specifically detects KCNN3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| KCNN3 | |
| Monoclonal | |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| hSK3, KCa2.3SKCA3small conductance calcium-activated potassium channel protein 3, potassium intermediate/small conductance calcium-activated channel, subfamilyN, member 3, SK3K3, SKCa 3, SKCa3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 631-731 of human KCNN3 (NP_002240.3).,, Sequence:, LSKMQNVMYDLITELNDRSEDLEKQIGSLESKLEHLTASFNSLPLLIADTLRQQQQQLLSAIIEARGVSVAVGTTHTPISDSPIGVSSTSFPTPYTSSSSC | |
| 20 μL | |
| Neuroscience, Signal Transduction | |
| 3782 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction