missing translation for 'onlineSavingsMsg'
Learn More

KCNN2 Rabbit anti-Human, Rat, Monkey, Polyclonal, Novus Biologicals™

Product Code. 18379490 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379490 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379490 Supplier Novus Biologicals Supplier No. NBP2841110.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KCNN2 Polyclonal specifically detects KCNN2 in Human, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNN2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias hSK2, KCa2.2SKCA2, potassium intermediate/small conductance calcium-activated channel, subfamilyN, member 2, SK2apamin-sensitive small-conductance Ca2+-activated potassium channel, SKCa 2, SKCa2, small conductance calcium-activated potassium channel protein 2
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C terminal region of human KCNN2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3781
Target Species Human, Rat, Monkey
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.