missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17929-1-AP-20UL
This item is not returnable.
View return policy
Description
Action potentials in vertebrate neurons are followed by an afterhyperpolarization (AHP) that may persist for several seconds and may have profound consequences for the firing pattern of the neuron. Each component of the AHP is kinetically distinct and is mediated by different calcium-activated potassium channels. The protein encoded by this gene is activated before membrane hyperpolarization and is thought to regulate neuronal excitability by contributing to the slow component of synaptic AHP. The encoded protein is an integral membrane protein that forms a voltage-independent calcium-activated channel with three other calmodulin-binding subunits. KCNN1 is a member of the KCNN family of potassium channels.Specifications
| KCNN1 | |
| Polyclonal | |
| Unconjugated | |
| KCNN1 | |
| hSK1, KCa2.1, KCNN1, SK, SK1, SKCa 1, SKCA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3780, 54261, 84036 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P70606, Q92952, Q9EQR3 | |
| KCNN1 | |
| KCNN1 Fusion Protein Ag12357 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction