missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNN1 Polyclonal Antibody

KCNN1 Polyclonal Antibody
Brand: Invitrogen PA597206
This item is not returnable.
View return policy
Description
KCNN1 Polyclonal Antibody for Western Blot
Specifications
| KCNN1 | |
| Polyclonal | |
| Unconjugated | |
| Kcnn1 | |
| AI854248; hSK1; KCa2.1; Kcnn1; potassium calcium-activated channel subfamily N member 1; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 1; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 1; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 1-like; SK; Sk1; SKCa 1; SKCa1; small conductance Ca2+-activated K+ channel type1; small conductance calcium-activated potassium channel 1; small conductance calcium-activated potassium channel protein 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3780, 84036 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.85 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q92952, Q9EQR3 | |
| Kcnn1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human KCNN1 (NP_0022392). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction