missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNMB4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550692
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total KCNMB4 protein.
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which slows activation kinetics, leads to steeper calcium sensitivity, and shifts the voltage range of current activation to more negative potentials than does the beta 1 subunit.
Specifications
| KCNMB4 | |
| Polyclonal | |
| Unconjugated | |
| KCNMB4 | |
| 1700058G18Rik; 2900045G12Rik; big potassium channel beta subunit 4; BK channel beta subunit 4; BK channel subunit beta-4; BKbeta4; calcium activated potassium channel beta 4 subunit; calcium-activated potassium channel subunit beta-4; Calcium-activated potassium channel, subfamily M subunit beta-4; charybdotoxin receptor subunit beta-4; hbeta4; k(VCA)beta-4; KCMB4; KCNMB4; large conductance calcium-dependent potassium ion channel beta 4 subunit; maxi K channel subunit beta-4; MaxiK channel beta-subunit 4; potassium calcium-activated channel subfamily M regulatory beta subunit 4; potassium channel subfamily M regulatory beta subunit 4; potassium large conductance calcium-activated channel, subfamily M, beta member 4; slo-beta-4; Slowpoke beta 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27345, 58802 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.9 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q86W47, Q9JIN6 | |
| KCNMB4 | |
| Synthetic peptide corresponding to residues near the N terminal of human potassium large conductance calcium-activated channel, subfamily M, beta member 4. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction