missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNMB4 Mouse anti-Human, Mouse, Clone: 4H7G3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60122-1-IG-20UL
This item is not returnable.
View return policy
Description
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which slows activation kinetics, leads to steeper calcium sensitivity, and shifts the voltage range of current activation to more negative potentials than does the beta 1 subunit.Specifications
| KCNMB4 | |
| Monoclonal | |
| 0.83 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86W47, Q9JIN6 | |
| Kcnmb4 | |
| KCNMB4 Fusion Protein Ag5618 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 4H7G3 | |
| Unconjugated | |
| KCNMB4 | |
| BK channel subunit beta 4, BKbeta4, Hbeta4, K(VCA)beta 4, KCNMB4, Maxi K channel subunit beta 4, Slo beta 4 | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 27345, 58802 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction