missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KCNMB3 Monoclonal antibody specifically detects KCNMB3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Microarray
Specifications
Specifications
| Antigen | KCNMB3 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Microarray |
| Classification | Monoclonal |
| Clone | S40B-18 |
| Conjugate | Janelia Fluor 549 |
| Dilution | Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Microarray |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BK channel subunit beta-3, BKbeta3, HBETA3, KCNMB2, KCNMBL, potassium large conductance calcium-activated channel, subfamily M beta member3, slo-beta-3, subfamily M subunit beta-3, voltage and Ca2+ activated potassium channel Maxi K beta 3subunit |
| Host Species | Mouse |
| Immunogen | Fusion protein amino acids 1-49 (unique N-terminus) of mouse BKBeta3a |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?