missing translation for 'onlineSavingsMsg'
Learn More

KCNMB3 Antibody (S40B-18), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18424713 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424713 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424713 Supplier Novus Biologicals Supplier No. NBP212916AF594

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KCNMB3 Monoclonal antibody specifically detects KCNMB3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Microarray
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNMB3
Applications Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Microarray
Classification Monoclonal
Clone S40B-18
Conjugate Alexa Fluor 594
Dilution Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Microarray
Formulation 50mM Sodium Borate
Gene Alias BK channel subunit beta-3, BKbeta3, HBETA3, KCNMB2, KCNMBL, potassium large conductance calcium-activated channel, subfamily M beta member3, slo-beta-3, subfamily M subunit beta-3, voltage and Ca2+ activated potassium channel Maxi K beta 3subunit
Host Species Mouse
Immunogen Fusion protein amino acids 1-49 (unique N-terminus) of mouse BKBeta3a
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 27094
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.