missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNMB3 Antibody (5H1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00027094-M02
This item is not returnable.
View return policy
Description
KCNMB3 Monoclonal antibody specifically detects KCNMB3 in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| KCNMB3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| BK channel subunit beta-3, BKbeta3, HBETA3, KCNMB2, KCNMBL, potassium large conductance calcium-activated channel, subfamily M beta member3, slo-beta-3, subfamily M subunit beta-3, voltage and Ca2+ activated potassium channel Maxi K beta 3subunit | |
| KCNMB3 (NP_741979, 82 a.a. ~ 181 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. FMLSIQREESTCTAIHTDIMDDWLDCAFTCGVHCHGQGKYPCLQVFVNLSHPGQKALLHYNEEAVQINPKCFYTPKCHQDRNDLLNSALDIKEFFDHKNG | |
| 0.1 mg | |
| Neuroscience | |
| 27094 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 5H1 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_741979 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction