missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNMB2 Antibody (S53-32), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12917-0.025mg
This item is not returnable.
View return policy
Description
KCNMB2 Monoclonal antibody specifically detects KCNMB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Microarray
Specifications
| KCNMB2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Microarray | |
| BK channel subunit beta-2, BKbeta2, calcium-activated potassium channel subunit beta-2, Calcium-activated potassium channel, subfamily M subunit beta-2, Charybdotoxin receptor subunit beta-2, hbeta2, Hbeta3, k(VCA)beta-2, large conductance calcium-activated potassium channel beta 2 subunit, large-conductance Ca2+-activated K+ channel beta2 subunit, Maxi K channel subunit beta-2, MaxiK channel beta 2 subunit, MGC22431, potassium large conductance calcium-activated channel, subfamily M, beta member2, slo-beta-2 | |
| Fusion protein amino acids 1-41 (N-terminus) and 218-235 (C-terminus) of mouse BKBeta2 | |
| 0.025 mg | |
| Neuroscience | |
| 10242 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| S53-32 | |
| Unconjugated | |
| PBS (pH 7.4), 50% Glycerol | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction