missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNMB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587724
This item is not returnable.
View return policy
Description
Immunogen sequence: TYYILVTTVL PLYQKSVWTQ ESKCHLIETN IRDQEELKGK KVPQYPCLWV NVSAAGRWAV LYHTEDTRDQ NQQ; Positive Samples: U-87MG, THP-1, BxPC-3; Cellular Location: Membrane, Multi-pass membrane protein.
Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open. Maxi K+ beta is the target for possible therapeutics because of its role in blood flow and blood pressure regulation.
Specifications
| KCNMB1 | |
| Polyclonal | |
| Unconjugated | |
| KCNMB1 | |
| big potassium channel beta subunit 1; BK channel beta subunit; BK channel beta subunit 1; BK channel subunit beta-1; BKbeta; BKbeta1; calcium-activated potassium channel beta subunit; calcium-activated potassium channel subunit beta; Calcium-activated potassium channel subunit beta-1; calcium-activated potassium channel, subfamily M subunit beta-1; Charybdotoxin receptor subunit beta-1; hbeta1; hslo-beta; K(VCA)beta; k(VCA)beta-1; Kcnmb1; large conductance Ca2+-activated K+ channel beta 1 subunit; maxi K channel subunit beta-1; MaxiK channel beta-subunit 1; potassium calcium-activated channel subfamily M regulatory beta subunit 1; potassium channel subfamily M regulatory beta subunit 1; potassium large conductance calcium-activated channel, subfamily M, beta member 1; slo-beta; slo-beta-1; slowpoke-beta | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3779 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16558 | |
| KCNMB1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-102 of human KCNMB1 (NP_0041281). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction