Learn More
KCNMA1 Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Brand: Invitrogen PA5111800
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). GST fusion protein sequence is: SHSSHSSQS SSKKSSSVHS IPSTANRPNR PKSRESRDKQ NATRMTRMGQ AEKKWFTDEP DNAYPRNIQI KPMSTHMANQ INQYKSTSSL IPPIREVEDE C Homology: Mouse - 21/21 amino acid residues identical to the canonical sequence (AccessionQ08460); rat - 20/21 amino acid residues identical to the canonical sequence (AccessionQ62976); human - 19/21 amino acid residues identical to the canonical sequence (AccessionQ12791).
Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.
Specifications
| KCNMA1 | |
| Polyclonal | |
| Unconjugated | |
| Kcnma1 | |
| 5730414M22Rik; bA205K10.1; BK channel; BKCa; BKCA alpha; BKTM; Calcium-activated potassium channel subunit alpha-1; Calcium-activated potassium channel, subfamily M subunit alpha-1; hSlo; K(VCA)alpha; KCa1.1; Kcnma; KCNMA1; KCNMA1b; KCNMA1c; Maxi K channel; MaxiK; Maxi-K channel HSLO; mSlo; mSlo1; SAKCA; SLO; Slo homolog; Slo1; Slo-alpha; Slowpoke homolog | |
| Guinea Pig | |
| Antigen affinity chromatography | |
| RUO | |
| 16531, 3778, 83731 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q08460, Q12791, Q62976 | |
| Kcnma1 | |
| GST fusion protein corresponding to residues 1097-1196 of mouse KCNMA1variant 2 (Intracellular, C-terminus) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.