missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNK6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559095
This item is not returnable.
View return policy
Description
Immunogen sequence: LQTFRHVSDL HGLTELILLP PPCPASFNAD EDDRVDILGP QPESHQQLSA SSHTDY Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 68%.
KCNK6 channels play a critical role in maintaining the electrical stability of cells, and their dysfunction can contribute to a variety of pathophysiological conditions. KCNK6, also known as TWIK-2 (Tandem of Pore Domains in a Weak Inwardly Rectifying K+ Channel 2), is a member of the two-pore-domain potassium (K2P) channel family.
Specifications
| KCNK6 | |
| Polyclonal | |
| Unconjugated | |
| KCNK6 | |
| D7Ertd764e; FLJ12282; inward rectifying potassium channel protein TWIK-2; K2p6.1; K2P6.1 potassium channel; Kcnk6; KCNK8; Potassium channel subfamily K member 6; potassium channel, subfamily K, member 6; potassium channel, two pore domain subfamily K, member 6; potassium inwardly-rectifying channel, subfamily K, member 6; potassium two pore domain channel subfamily K member 6; TOSS; TWIK2; TWIK-2; TWIK-2 two-pore-domain K+ channel; TWIK-originated similarity sequence; TWIK-originated sodium similarity sequence | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9424 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y257 | |
| KCNK6 | |
| Recombinant protein corresponding to Human KCNK6. Recombinant protein control fragment (Product #RP-98354). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur