missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNK6 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA541330

Product Code. 15574772

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Peptide sequence: RLGRVVLANA SGSANASDPA WDFASALFFA STLITTVGYG YTTPLTDAGK Sequence homology: Cow: 100%; Human: 100%; Pig: 100%.

KCNK6 channels play a critical role in maintaining the electrical stability of cells, and their dysfunction can contribute to a variety of pathophysiological conditions. KCNK6, also known as TWIK-2 (Tandem of Pore Domains in a Weak Inwardly Rectifying K+ Channel 2), is a member of the two-pore-domain potassium (K2P) channel family.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KCNK6
Polyclonal
Unconjugated
KCNK6
D7Ertd764e; FLJ12282; inward rectifying potassium channel protein TWIK-2; K2p6.1; K2P6.1 potassium channel; Kcnk6; KCNK8; Potassium channel subfamily K member 6; potassium channel, subfamily K, member 6; potassium channel, two pore domain subfamily K, member 6; potassium inwardly-rectifying channel, subfamily K, member 6; potassium two pore domain channel subfamily K member 6; TOSS; TWIK2; TWIK-2; TWIK-2 two-pore-domain K+ channel; TWIK-originated similarity sequence; TWIK-originated sodium similarity sequence
Rabbit
Affinity Chromatography
RUO
9424
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9Y257
KCNK6
Synthetic peptide directed towards the N-terminal of human KCNK6 (aa 71-120).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.