missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577598
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
KCNK5 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. KCNK5 is mainly expressed in the cortical distal tubules and collecting ducts of the kidney. The protein is highly sensitive to external pH and this, in combination with its expression pattern, suggests it may play an important role in renal potassium transport.
Specifications
| KCNK5 | |
| Polyclonal | |
| Unconjugated | |
| KCNK5 | |
| TASK-2, Kcnk5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16529, 364241, 8645 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 1% BSA, 5% sucrose and 0.05% sodium azide; pH 7.4 | |
| O95279 | |
| KCNK5 | |
| Peptide (C)YEQLMNEYNKANSPKGT, amino acid residues 483-499 of human K2P 5.1 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction