missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12615-50ul
This item is not returnable.
View return policy
Description
KCNK5 Polyclonal antibody specifically detects KCNK5 in Human samples. It is validated for Western Blot
Specifications
| KCNK5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Acid-sensitive potassium channel protein TASK-2, FLJ11035, K2p5.1, potassium channel subfamily K member 5, potassium channel, subfamily K, member 1 (TASK-2), potassium channel, subfamily K, member 5, TASK-2, TASK2K2P5.1 potassium channel, TWIK-related acid-sensitive K(+) channel 2, TWIK-related acid-sensitive K+ channel 2 | |
| Produced in rabbits immunized with E. coli-derived Human KCNK5 fragment | |
| 50 μg | |
| Signal Transduction | |
| 8645 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction