missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | KCNK4 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18382457
|
Novus Biologicals
NBP3-04902-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18352216
|
Novus Biologicals
NBP3-04902-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KCNK4 Polyclonal specifically detects KCNK4 in Human samples. It is validated for Western Blot.Specifications
| KCNK4 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 50801 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| K2p4.1, K2P4.1 potassium channel, potassium channel subfamily K member 4, potassium channel, subfamily K, member 4, TRAAKTRAAK1, TWIK-related arachidonic acid-stimulated potassium channel protein, Two pore K(+) channel KT4.1, two pore K+ channel KT4.1, Two pore potassium channel KT4.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-90 of human KCNK4 (NP_201567.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title