missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27113-1-AP-20UL
This item is not returnable.
View return policy
Description
Potassium channels play a role in many cellular processes including maintece of the action potential, muscle contraction, hormone secretion, osmotic regulation, and ion flow. This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The encoded protein homodimerizes and functions as an outwardly rectifying channel. It is expressed primarily in neural tissues and is stimulated by membrane stretch and polyunsaturated fatty acids.Specifications
| KCNK4 | |
| Polyclonal | |
| Unconjugated | |
| KCNK4 | |
| K2p4.1, KCNK4, TRAAK, TRAAK1, Two pore K(+) channel KT4.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116489, 16528, 50801 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| G3V8V5, O88454, Q9NYG8 | |
| KCNK4 | |
| KCNK4 Fusion Protein Ag25931 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction