missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK18 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23712-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can be used to detect endogenous KCNK18, however, the reason for the additional band of 130-140 kDa is still unknown.
TRESK (TWIK-related spinal cord K+ channel) is a two-pore domain K+ channel that induces outward rectification and functions as a background K+ channel that is abundantly expressed in the spinal cord, thymus, spleen, and leukemic T-lymphocytes. TRESK is activated by increased cytoplasmic calcium concentration through calcineurin as well as by acetylcholine and histamine. It has been suggested that TRESK is linked to acute and chronic pain by activation of calcineurin. Other studies indicate that TRESK may play a role in the regulation of T-cell function and could be an excellent target to treat T-cell originated immune dysfunction.Specifications
| KCNK18 | |
| Polyclonal | |
| Unconjugated | |
| KCNK18 | |
| K2p18.1, KCNK18, TRESK, TRESK 2, TRESK2, TRIK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 338567 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z418 | |
| KCNK18 | |
| KCNK18 Fusion Protein Ag20514 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction