missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNK18 Polyclonal Antibody

Product Code. 12839022
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12839022 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12839022 Supplier Invitrogen™ Supplier No. PA520873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-20873 can be used with blocking peptide PEP-0987.

TRESK (TWIK-related spinal cord K+ channel) is a two-pore domain K+ channel that induces outward rectification and functions as a background K+ channel that is abundantly expressed in the spinal cord, thymus, spleen, and leukemic T-lymphocytes. TRESK is activated by increased cytoplasmic calcium concentration through calcineurin as well as by acetylcholine and histamine. It has been suggested that TRESK is linked to acute and chronic pain by activation of calcineurin. Other studies indicate that TRESK may play a role in the regulation of T-cell function and could be an excellent target to treat T-cell originated immune dysfunction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK18
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene KCNK18
Gene Accession No. Q6Q1P3, Q6VV64, Q7Z418
Gene Alias 2P K ion channel TRESK; Gm781; K2p18.1; Kcnk18; MGR13; Potassium channel subfamily K member 18; potassium channel, subfamily K, member 18; potassium channel, two pore domain subfamily K, member 18; potassium two pore domain channel subfamily K member 18; tandem-pore K+ (K(2P)) channel; Tresk; TRESK2; Tresk-2; TRIK; TWIK-related individual K+ channel; TWIK-related individual potassium channel; TWIK-related spinal cord K+ channel; TWIK-related spinal cord potassium channel; Two-pore-domain potassium channel TRESK
Gene Symbols KCNK18
Host Species Rabbit
Immunogen A 17 amino acid peptide from near the center of human TRESK.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 332396, 338567, 445371
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.