missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539345
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the family of potassium channel proteins containing two pore-forming P domains. This channel is an open rectifier which primarily passes outward current under physiological K+ concentrations. This gene is activated at alkaline pH. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2008].
Specifications
| KCNK17 | |
| Polyclonal | |
| Unconjugated | |
| KCNK17 | |
| TALK2, TASK4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 89822 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96T54 | |
| KCNK17 | |
| A synthetic peptide derived from the internal region of human KCNK17 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction