Learn More
Invitrogen™ KCNK13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113607
Description
Immunogen sequence: GEMISMKDLL AANKASLAIL QKQLSEMANG CPHQTSTLAR DNEFSGGVGA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Horse: 79%; Human: 100%; Pig: 79%; Rat: 79%.
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a potassium channel containing two pore-forming domains. This protein is an open channel that can be stimulated by arachidonic acid and inhibited by the anesthetic halothane.
Specifications
| KCNK13 | |
| Polyclonal | |
| Unconjugated | |
| KCNK13 | |
| BB085247; F730021E22Rik; Gm1570; Gm1685; K2p13.1; K2P13.1 potassium channel; KCNK13; Potassium channel subfamily K member 13; potassium channel, subfamily K, member 13; potassium channel, two pore domain subfamily K, member 13; potassium two pore domain channel subfamily K member 13; prdx1; Tandem pore domain halothane-inhibited potassium channel 1; tandem pore domain potassium channel THIK-1; THIK1; THIK-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56659 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9HB14 | |
| KCNK13 | |
| Synthetic peptide directed towards the C terminal region of human KCNK13 (aa 344-393). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.