missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNK13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572686
This item is not returnable.
View return policy
Description
KCNK13 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a potassium channel containing two pore-forming domains. This protein is an open channel that can be stimulated by arachidonic acid and inhibited by the anesthetic halothane.
Specifications
| KCNK13 | |
| Polyclonal | |
| Unconjugated | |
| KCNK13 | |
| BB085247; F730021E22Rik; Gm1570; Gm1685; K2p13.1; K2P13.1 potassium channel; KCNK13; Potassium channel subfamily K member 13; potassium channel, subfamily K, member 13; potassium channel, two pore domain subfamily K, member 13; potassium two pore domain channel subfamily K member 13; prdx1; Tandem pore domain halothane-inhibited potassium channel 1; tandem pore domain potassium channel THIK-1; THIK1; THIK-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 217826, 56659, 64120 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8R1P5, Q9ERS0, Q9HB14 | |
| KCNK13 | |
| KCNK13 antibody for a 15 amino acid synthetic peptide near the center of human KCNK13. The immunogen is within amino acids 150 - 200 of KCNK13. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering