missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals KCNK13 Antibody Blocking Peptide

Product Code. 18691666 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18691666

Brand: Novus Biologicals NBP241132PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene ID (Entrez) 56659
Species Human
Accession Number NP_071337
Concentration 0.2 mg/mL
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
For Use With (Application) KCNK13 peptide is used for blocking the activity of KCNK13 antibody NBP2-41132.
Gene Alias K2p13.1, potassium channel subfamily K member 13, potassium channel, subfamily K, member 13, tandem pore domain potassium channel THIK-1, THIK-1
Gene Symbol KCNK13
Quantity 0.05 mg
Research Category Signal Transduction
Storage Buffer PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.