missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KCNK12 Polyclonal antibody specifically detects KCNK12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | KCNK12 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Research Discipline | Signal Transduction |
| Primary or Secondary | Primary |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?