missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK10 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | KCNK10 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18367884
|
Novus Biologicals
NBP3-05006-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18383576
|
Novus Biologicals
NBP3-05006-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KCNK10 Polyclonal specifically detects KCNK10 in Human samples. It is validated for Western Blot.Specifications
| KCNK10 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 54207 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 2P domain potassium channel TREK2, K2p10.1, Outward rectifying potassium channel protein TREK-2, potassium channel subfamily K member 10, potassium channel TREK-2, potassium channel, subfamily K, member 10, TREK-2, TREK-2 K(+) channel subunit, TREK2FLJ43399, TWIK-related K+ channel 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 384-543 of human KCNK10 (NP_612191.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title