missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNK10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115451
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KCNK10.
KCNK10 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The message for this gene is highly expressed in the kidney and pancreas. This channel is an open rectifier which primarily passes outward current under physiological K+ concentrations. The protein is stimulated strongly by arachidonic acid and to a lesser degree by membrane stretching, intracellular acidification, and general anaesthetics. Three transcript variants have been identified for this gene.
Specifications
| KCNK10 | |
| Polyclonal | |
| Unconjugated | |
| KCNK10 | |
| 1700024D23Rik; 2P domain potassium channel TREK2; 3010005K24Rik; K2p10.1; Kcnk10; Outward rectifying potassium channel protein TREK-2; outward rectifying potassium channel TREK2; potassium channel subfamily K member 10; potassium channel TREK-2; potassium channel, subfamily K, member 10; potassium channel, two pore domain subfamily K, member 10; potassium two pore domain channel subfamily K member 10; PPP1R97; protein phosphatase 1, regulatory subunit 97; TREK2; TREK-2; TREK-2 K(+) channel subunit; TREK-2 two-pore-domain K+ channel; TWIK-related K+ channel 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54207, 65272, 72258 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P57789, Q9JIS4 | |
| KCNK10 | |
| A synthesized peptide derived from human KCNK10(Accession P57789), corresponding to amino acid residues R456-D506. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction