missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-87664
This item is not returnable.
View return policy
Description
KCNK10 Polyclonal specifically detects KCNK10 in Human samples. It is validated for Western Blot.
Specifications
| KCNK10 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54207 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| 2P domain potassium channel TREK2, K2p10.1, Outward rectifying potassium channel protein TREK-2, potassium channel subfamily K member 10, potassium channel TREK-2, potassium channel, subfamily K, member 10, TREK-2, TREK-2 K(+) channel subunit, TREK2FLJ43399, TWIK-related K+ channel 2 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of human KCNK10 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction