missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41301R
This item is not returnable.
View return policy
Description
KCNK1 Polyclonal antibody specifically detects KCNK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| KCNK1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| DPK, HOHO1, Inward rectifying potassium channel protein TWIK-1, K2P1, K2p1.1, KCNO1, Potassium channel KCNO1, potassium channel subfamily K member 1, potassium channel, subfamily K, member 1, potassium inwardly-rectifying channel, subfamily K, member 1, TWIK-1, TWIK1HOHO | |
| Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human KCNK1. The immunogen is located within amino acids 220 - 270 of KCNK1. Amino Acid Squence: DYVPGEGYNQKFRELYK | |
| 0.1 mL | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 3775 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction