missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kcnj8, Mouse anti-Rat, Clone: S366-60, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16466.100ug
This item is not returnable.
View return policy
Specifications
| Kcnj8 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
| Kir6.1/UKATP1/uKATP-1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S366-60 | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| potassium inwardly-rectifying channel, subfamily J, member 8 | |
| Kcnj8 | |
| Recombinant protein corresponding to amino acids 306-424 at C-terminus of rat Kcnj8. | |
| RUO | |
| 25472 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction