missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNJ13 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£163.00 - £380.00
Specifications
| Antigen | KCNJ13 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18321997
|
Novus Biologicals
NBP3-04751-20UL |
20 μL |
£163.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18396196
|
Novus Biologicals
NBP3-04751-100UL |
100 μL |
£380.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KCNJ13 Polyclonal specifically detects KCNJ13 in Rat samples. It is validated for Western Blot.Specifications
| KCNJ13 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| Inward rectifier K(+) channel Kir7.1, inwardly rectifying subfamily J member 13, MGC33328, potassium inwardly-rectifying channel, subfamily J, member 13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-360 of human KCNJ13 (NP_002233.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS with 50% glycerol, pH7.3. | |
| 3769 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title