Learn More
KCNJ10 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP1-70371-0.025ML
 View more versions of this product
Product Code. 18064724
  • £163.00 / 25µL
Description
KCNJ10 Polyclonal specifically detects KCNJ10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KCNJ10
Polyclonal
Unconjugated
Whole antisera with No Preservative
KCNJ10
A synthetic peptide from aa region 330-379 of human KCNJ10 conjugated to an immunogenic carrier protein was used as the antigen. The peptide is homologous in mouse and rat.
0.025 mL
Primary
Rat, mouse, human. Other species not yet tested.
Mouse, Human, Rat
Antisera
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
LYOPH
Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
ATP-dependent inwardly rectifying potassium channel Kir4.1, ATP-sensitive inward rectifier potassium channel 10, BIRK-10, glial ATP-dependent inwardly rectifying potassium channel KIR4.1, Inward rectifier K(+) channel Kir1.2, inward rectifier K+ channel KIR1.2, KCNJ13-PEN, KIR1.2, Kir4.1, Potassium channel, inwardly rectifying subfamily J member 10, potassium inwardly-rectifying channel, subfamily J, member 10, SESAME
Goat
Unpurified
RUO
3766
Reconstitute with 50 ul sterilized water to desired concetration.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.