missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNJ10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £397.00
Specifications
| Antigen | KCNJ10 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18248434
|
Novus Biologicals
NBP1-20149-0.025ML |
0.025 mL |
£153.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18748743
|
Novus Biologicals
NBP1-20149 |
£397.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
KCNJ10 Polyclonal specifically detects KCNJ10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| KCNJ10 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| ATP-dependent inwardly rectifying potassium channel Kir4.1, ATP-sensitive inward rectifier potassium channel 10, BIRK-10, glial ATP-dependent inwardly rectifying potassium channel KIR4.1, Inward rectifier K(+) channel Kir1.2, inward rectifier K+ channel KIR1.2, KCNJ13-PEN, KIR1.2, Kir4.1, Potassium channel, inwardly rectifying subfamily J member 10, potassium inwardly-rectifying channel, subfamily J, member 10, SESAME | |
| KCNJ10 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| P78508 | |
| 3766 | |
| IgG | |
| Unpurified | |
| KCNJ10. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title