missing translation for 'onlineSavingsMsg'
Learn More

KCNJ10 Antibody (1C11), Novus Biologicals™

Product Code. 18377888 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377888 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377888 Supplier Novus Biologicals Supplier No. H00003766M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KCNJ10 Monoclonal antibody specifically detects KCNJ10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNJ10
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 1C11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunoprecipitation, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002232
Gene Alias ATP-dependent inwardly rectifying potassium channel Kir4.1, ATP-sensitive inward rectifier potassium channel 10, BIRK-10, glial ATP-dependent inwardly rectifying potassium channel KIR4.1, Inward rectifier K(+) channel Kir1.2, inward rectifier K+ channel KIR1.2, KCNJ13-PEN, KIR1.2, Kir4.1, Potassium channel, inwardly rectifying subfamily J member 10, potassium inwardly-rectifying channel, subfamily J, member 10, SESAME
Host Species Mouse
Immunogen KCNJ10 (NP_002232, 276 a.a. ~ 379 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DFELVLILSGTVESTSATCQVRTSYLPEEILWGYEFTPAISLSASGKYIADFSLFDQVVKVASPSGLRDSTVRYGDPEKLKLEESLREQAEKEGSALSVRISNV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3766
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.