missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNIP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540940
This item is not returnable.
View return policy
Description
Peptide sequence: NSTKRSIKER LMKLLPCSAA KTSSPAIQNS VEDELEMATV RHRPEALELL Sequence homology: Cow: 93%; Dog: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
KCNIP4 encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. This protein member also interacts with presenilin.
Specifications
| KCNIP4 | |
| Polyclonal | |
| Unconjugated | |
| KCNIP4 | |
| A-type potassium channel modulatory protein 4; AV032399; Calp; Calp250; calsenilin-like protein; KChIP4; KChIP4a; KCNIP4; Kv channel interacting protein 4; Kv channel-interacting protein 4; potassium channel interacting protein 4; potassium channel-interacting protein 4; potassium voltage-gated channel interacting protein 4; Unknown (protein for MGC:137737) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80333 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6PIL6 | |
| KCNIP4 | |
| Synthetic peptide directed towards the N-terminal of human KCNIP4 (aa 27-76). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction