missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141705
This item is not returnable.
View return policy
Description
Suggested positive control: human heart and brain protein or tissue. Immunogen sequence: LQTKQRRPSK DKIEDELE.
This gene encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. Alternative splicing results in multiple transcript variant encoding different isoforms.
Specifications
| KCNIP1 | |
| Polyclonal | |
| Unconjugated | |
| Kcnip1 | |
| ABP; A-type potassium channel modulatory protein 1; A-type potassium channel modulatory protein 1.2; KChIP1; Kchip1.2; KCHIP2; Kcnip1; Kv channel interacting protein 1; Kv channel-interacting protein 1; potassium channel interacting protein 1; potassium channel-interacting protein 1; potassium voltage-gated channel interacting protein 1; VABP; Vesicle APC-binding protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 30820, 541458, 595102, 65023, 70357 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.2% gelatin and 0.05% sodium azide | |
| Q8R426, Q9JJ57, Q9NZI2 | |
| Kcnip1 | |
| Within the range of amino acids 1-35 of human ABP protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction