missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNH7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577627
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit. There are at least two alternatively spliced transcript variants derived from this gene and encoding distinct isoforms.
Specifications
| KCNH7 | |
| Polyclonal | |
| Unconjugated | |
| KCNH7 | |
| KCNH7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 170738, 170739, 90134 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| 0.6 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| O54852, Q9ER47, Q9NS40 | |
| Kcnh7 | |
| Peptide CPEFLDLEKSKLKSKE, corresponding to amino acid residues 1108-1123 of rat Kv 11.3 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction