missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNH7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118914
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KCNH7.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit. There are at least two alternatively spliced transcript variants derived from this gene and encoding distinct isoforms.
Specifications
| KCNH7 | |
| Polyclonal | |
| Unconjugated | |
| KCNH7 | |
| 9330137I11Rik; eag-related protein 3; erg3; ERG-3; ether-a-go-go-related gene potassium channel 3; ether-a-go-go-related protein 3; H Kv11.3; HERG3; KCNH7; Kv11.3; potassium channel erg3; potassium channel protein erg3; potassium channel subunit HERG-3; potassium channel, voltage gated eag related subfamily H, member 7; potassium voltage-gated channel subfamily H member 7; potassium voltage-gated channel, subfamily H (eag-related), member 7; voltage-gated potassium channel subunit Kv11.3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170738, 170739, 90134 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O54852, Q9ER47, Q9NS40 | |
| KCNH7 | |
| A synthesized peptide derived from human KCNH7(Accession Q9NS40), corresponding to amino acid residues P1093-L1143. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction